accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 166964-35-8

IUPAC Name: 4-bromo-5-chlorothiophene-2-sulfonyl chloride
Synonyms: 4-bromo-5-chlorothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 295.97
Molecular Formula: C4HBrCl2O2S2
InChi Key: PABOKJJDABSQCK-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C=C(S1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results