accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 166964-36-9

IUPAC Name: 4-bromo-2,5-dichlorothiophene-3-sulfonyl chloride
Synonyms: 4-bromo-2,5-dichlorothiophene-3-sulfonyl chloride
Molecular Weight (g/mol): 330.42
Molecular Formula: C4BrCl3O2S2
InChi Key: NAHZODWKWDZKSZ-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C(=C(Cl)S1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results