accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 1670-46-8

IUPAC Name: 2-acetylcyclopentan-1-one
Synonyms: acetylcyclopentanone
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: OSWDNIFICGLKEE-UHFFFAOYNA-N
SMILES: CC(=O)C1CCCC1=O
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)