accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHN

CAS RN 1670-81-1

IUPAC Name: 1H-indole-5-carboxylate
Synonyms: 1H-indole-5-carboxylate
Molecular Weight (g/mol): 160.15
Molecular Formula: C9H6NO2
InChi Key: IENZCGNHSIMFJE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2NC=CC2=C1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)