accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16703-52-9

IUPAC Name: ethyl (dimethylcarbamoyl)formate
Synonyms: ethyl (dimethylcarbamoyl)formate
Molecular Weight (g/mol): 145.16
Molecular Formula: C6H11NO3
InChi Key: HMALWDVRMHVUAW-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)N(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results