accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 167393-62-6

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid
Molecular Weight (g/mol): 624.78
Molecular Formula: C41H40N2O4
InChi Key: YPTNAIDIXCOZAJ-WHUAGQPENA-N
SMILES: CC1=CC=C(C=C1)C(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)