accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16741-80-3

HClH3COONH2S

CAS RN 16741-80-3

IUPAC Name: hydrogen methyl 2-amino-3-(benzylsulfanyl)propanoate chloride
Synonyms: hydrogen methyl 2-amino-3-(benzylsulfanyl)propanoate chloride
Molecular Weight (g/mol): 261.76
Molecular Formula: C11H16ClNO2S
InChi Key: QVJDVOZRQMIIHP-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(N)CSCC1=CC=CC=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)