accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16744-98-2

IUPAC Name: 1-fluoro-2-isocyanatobenzene
Synonyms: 1-fluoro-2-isocyanatobenzene
Molecular Weight (g/mol): 137.11
Molecular Formula: C7H4FNO
InChi Key: VZNCSZQPNIEEMN-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1N=C=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)