accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3CH3H3C

CAS RN 16747-26-5

IUPAC Name: 2,2,4-trimethylhexane
Synonyms: 2,2,4-trimethylhexane
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: AFTPEBDOGXRMNQ-UHFFFAOYNA-N
SMILES: CCC(C)CC(C)(C)C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)