CAS RN 167479-16-5
CAS RN 167479-16-5
IUPAC Name:
1-methyl-1H-indole-7-carboxylic acid
Synonyms:
1-methylindole-7-carboxylic acid
Molecular Weight (g/mol):
175.19
Molecular Formula:
C10H9NO2
InChi Key:
DWKIPLIDZYRILV-UHFFFAOYSA-N
SMILES:
CN1C=CC2=CC=CC(C(O)=O)=C12
1
–
1
of
1
results
1
1-Methyl-1H-indole-7-carboxylic acid, 97%, Thermo Scientific™
CAS: 167479-16-5 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.19 MDL Number: MFCD09879957 InChI Key: DWKIPLIDZYRILV-UHFFFAOYSA-N PubChem CID: 18388362 IUPAC Name: 1-methylindole-7-carboxylic acid SMILES: CN1C=CC2=CC=CC(C(O)=O)=C12