accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 167487-83-4

CH3OOHNN

CAS RN 167487-83-4

IUPAC Name: ethyl 1H-1,3-benzodiazole-4-carboxylate
Synonyms: ethyl 1H-1,3-benzodiazole-4-carboxylate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: BIROFVBTDWNLKU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2N=CNC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results