accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16753-62-1

IUPAC Name: ethenyldimethoxymethylsilane
Synonyms: ethenyldimethoxymethylsilane
Molecular Weight (g/mol): 132.23
Molecular Formula: C5H12O2Si
InChi Key: ZLNAFSPCNATQPQ-UHFFFAOYSA-N
SMILES: CO[Si](C)(OC)C=C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)