accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3O

CAS RN 1676-63-7

IUPAC Name: 1-(4-ethoxyphenyl)ethan-1-one
Synonyms: 1-(4-ethoxyphenyl)ethanone
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: YJFNFQHMQJCPRG-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C(C)=O
Boiling Point 
  • (3)
  • (2)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (1)
  • (5)
  • (4)
Quantity 
  • (3)
  • (4)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (1)
  • (7)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (8)
  • (1)
  • (5)
  • (4)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)