accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHNOOOHO

CAS RN 1676-75-1

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-(tert-butoxy)propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-(tert-butoxy)propanoic acid
Molecular Weight (g/mol): 295.34
Molecular Formula: C15H21NO5
InChi Key: TXDGEONUWGOCJG-UHFFFAOYNA-N
SMILES: CC(C)(C)OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results