accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOOO

CAS RN 167612-35-3

IUPAC Name: 3,4,5-tris(benzyloxy)-2-methyl-6-(phenylsulfanyl)oxane
Synonyms: 3,4,5-tris(benzyloxy)-2-methyl-6-(phenylsulfanyl)oxane
Molecular Weight (g/mol): 526.69
Molecular Formula: C33H34O4S
InChi Key: WGAOZACWMVPQOY-UHFFFAOYNA-N
SMILES: CC1OC(SC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)