accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 167692-95-7

IUPAC Name: 2-(2-cyclohexylethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2-cyclohexylethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 238.18
Molecular Formula: C14H27BO2
InChi Key: LDKDGYZAIIKCGK-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCC2CCCCC2)OC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results