accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1677-46-9

IUPAC Name: 2-hydroxy-1-methyl-1,4-dihydroquinolin-4-one
Synonyms: 2-hydroxy-1-methylquinolin-4-one
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: VWRAQWACTKTNAO-UHFFFAOYSA-N
SMILES: CN1C(O)=CC(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)