accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOHClNOO

CAS RN 16773-42-5

IUPAC Name: 1-chloro-3-(2-methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol
Synonyms: madelen
Molecular Weight (g/mol): 219.63
Molecular Formula: C7H10ClN3O3
InChi Key: IPWKIXLWTCNBKN-UHFFFAOYNA-N
SMILES: CC1=NC=C(N1CC(O)CCl)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)