accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1678-49-5

IUPAC Name: 3-bromo-4-nitropyridin-1-ium-1-olate
Synonyms: 3-bromo-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol): 218.99
Molecular Formula: C5H3BrN2O3
InChi Key: YZQMKADIENBVLH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=[N+]([O-])C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results