accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3

CAS RN 1678-92-8

IUPAC Name: propylcyclohexane
Synonyms: N-propylcyclohexane
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: DEDZSLCZHWTGOR-UHFFFAOYSA-N
SMILES: CCCC1CCCCC1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)