accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16789-84-7

IUPAC Name: 3-bromo-2-hydroxy-5-nitrobenzaldehyde
Synonyms: 3-bromo-2-hydroxy-5-nitrobenzaldehyde
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: BESBCGANGAEHPM-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)