accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1679-36-3

IUPAC Name: hex-3-yn-2-one
Synonyms: hex-3-yn-2-one
Molecular Weight (g/mol): 96.13
Molecular Formula: C6H8O
InChi Key: LTAPKZGQTMVYMX-UHFFFAOYSA-N
SMILES: CCC#CC(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)