accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1679-47-6

IUPAC Name: 3-methyloxolan-2-one
Synonyms: 3-methylbutyrolactone
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: QGLBZNZGBLRJGS-UHFFFAOYNA-N
SMILES: CC1CCOC1=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results