CAS RN 16793-91-2
IUPAC Name:
1-(2-bromoethyl)-2-chlorobenzene
Synonyms:
1-(2-bromoethyl)-2-chlorobenzene
Molecular Weight (g/mol):
219.51
Molecular Formula:
C8H8BrCl
InChi Key:
AECBVDLERUETKG-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1CCBr
1
–
1
of
1
results
1
Promotions Available
2-Chlorophenethyl bromide, 95%
2-Chlorophenethyl bromide, 95%
CAS: 16793-91-2 Molecular Formula: C8H8BrCl Molecular Weight (g/mol): 219.51 InChI Key: AECBVDLERUETKG-UHFFFAOYSA-N PubChem CID: 2756963 IUPAC Name: 1-(2-bromoethyl)-2-chlorobenzene SMILES: C1=CC=C(C(=C1)CCBr)Cl