accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOOCH3H3CCH3HO

CAS RN 167938-56-9

IUPAC Name: tert-butyl N-(2-hydroxypropyl)carbamate
Synonyms: tert-butyl N-(2-hydroxypropyl)carbamate
Molecular Weight (g/mol): 175.23
Molecular Formula: C8H17NO3
InChi Key: YNJCFDAODGKHAV-UHFFFAOYNA-N
SMILES: CC(O)CNC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)