accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrNCO

CAS RN 167951-51-1

IUPAC Name: 3-bromo-5-isocyanatopyridine
Synonyms: 3-bromo-5-isocyanatopyridine
Molecular Weight (g/mol): 199.01
Molecular Formula: C6H3BrN2O
InChi Key: PNEYUZGELZQGCX-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(=C1)N=C=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results