accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 1680-36-0

IUPAC Name: nonanoyl nonanoate
Synonyms: nonanoyl nonanoate
Molecular Weight (g/mol): 298.47
Molecular Formula: C18H34O3
InChi Key: PBBAFLCORNAZCD-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)OC(=O)CCCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)