accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 168034-31-9

CH3HOHNOOH3CCH3H3CO

CAS RN 168034-31-9

IUPAC Name: tert-butyl N-[3-(benzyloxy)-1-hydroxybutan-2-yl]carbamate
Synonyms: tert-butyl N-[3-(benzyloxy)-1-hydroxybutan-2-yl]carbamate
Molecular Weight (g/mol): 295.38
Molecular Formula: C16H25NO4
InChi Key: RXDBGDZAKNELGW-UHFFFAOYNA-N
SMILES: CC(OCC1=CC=CC=C1)C(CO)NC(=O)OC(C)(C)C
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)