accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaH3CCH2OO

CAS RN 16809-88-4

IUPAC Name: calcium 2-methylprop-2-enoate
Synonyms: calcium methacrylate
Molecular Weight (g/mol): 125.16
Molecular Formula: C4H5CaO2
InChi Key: ZUYRDYUKACMIRN-UHFFFAOYSA-M
SMILES: [Ca++].CC(=C)C([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results