accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 168106-22-7

IUPAC Name: bis(1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide) barium
Synonyms: bis(1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide) barium
Molecular Weight (g/mol): 699.61
Molecular Formula: C4H2BaF12N2O8S4
InChi Key: VJUKAFJLZPVWME-UHFFFAOYSA-N
SMILES: [Ba].FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results