accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3HO(R)OHONHONHSSHNOHNO(R)OHH3CCH3OH

CAS RN 16816-67-4

IUPAC Name: (2R)-N-[2-({2-[(2-{3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanamido}ethyl)disulfanyl]ethyl}carbamoyl)ethyl]-2,4-dihydroxy-3,3-dimethylbutanamide
Synonyms: pantethine pantomin D-pantethine pantetina bis(pantothenamidoethyl) disulfide
Molecular Weight (g/mol): 554.72
Molecular Formula: C22H42N4O8S2
InChi Key: DJWYOLJPSHDSAL-ROUUACIJSA-N
SMILES: CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO
Grade 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)