accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16827-96-6

IUPAC Name: λ²-cobalt(2+) ammonium hydrate phosphate
Synonyms: λ²-cobalt(2+) ammonium hydrate phosphate
Molecular Weight (g/mol): 189.96
Molecular Formula: CoH6NO5P
InChi Key: PNSIYUITZYKGRY-UHFFFAOYSA-L
SMILES: [NH4+].O.[Co++].[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results