accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 168300-88-7

IUPAC Name: (2S)-3-[(acetamidomethyl)sulfanyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-[(acetamidomethyl)sulfanyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 414.48
Molecular Formula: C21H22N2O5S
InChi Key: CSMYOORPUGPKAP-LJQANCHMSA-N
SMILES: CC(=O)NCSC[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)