accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16839-13-7

IUPAC Name: (4-bromophenyl)phosphonic acid
Synonyms: 4-bromophenylphosphonic acid
Molecular Weight (g/mol): 236.99
Molecular Formula: C6H6BrO3P
InChi Key: XDGIQCFWQNHSMV-UHFFFAOYSA-N
SMILES: OP(O)(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results