accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 1684-14-6

IUPAC Name: 2,3-diphenylquinoxaline
Synonyms: 2,3-diphenylquinoxaline
Molecular Weight (g/mol): 282.35
Molecular Formula: C20H14N2
InChi Key: RSNQVABHABAKEZ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C(N=C2C=CC=CC2=N1)C1=CC=CC=C1
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)