accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16846-24-5

IUPAC Name: (2S,3S,4R,6S)-6-{[(2R,3S,4R,5R,6S)-6-{[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-(acetyloxy)-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-6-yl]oxy}-4-(dimethylamino)-5-hydroxy-2-methyloxan-3-yl]oxy}-4-hydroxy-2,4-dimethyloxan-3-yl 3-methylbutanoate
Synonyms: josamycin josamycine josamycine [inn-french] josamycinum antibiotic yl-704 A3 交沙霉素 leucomycin A3 wilprafen josamicina [inn-spanish] turimycin A5
Molecular Weight (g/mol): 828.01
Molecular Formula: C42H69NO15
InChi Key: XJSFLOJWULLJQS-NGVXBBESSA-N
SMILES: CO[C@H]1[C@@H](CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]1O[C@H](C)[C@@H](O[C@H]2C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O2)[C@@H]([C@H]1O)N(C)C)OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results