accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16881-71-3

IUPAC Name: 4'-methoxy-[1,1'-biphenyl]-4-ol
Synonyms: 4'-methoxy-[1,1'-biphenyl]-4-ol
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: CORJIEYQXMZUIW-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results