accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONClO

CAS RN 16883-16-2

IUPAC Name: 5-methyl-3-phenyl-1,2-oxazole-4-carbonyl chloride
Synonyms: 5-methyl-3-phenyl-1,2-oxazole-4-carbonyl chloride
Molecular Weight (g/mol): 221.64
Molecular Formula: C11H8ClNO2
InChi Key: HXEVQMXCHCDPSO-UHFFFAOYSA-N
SMILES: CC1=C(C(Cl)=O)C(=NO1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results