accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3OOH

CAS RN 168899-58-9

IUPAC Name: 3-(acetyloxy)-2-methylbenzoic acid
Synonyms: 3-(acetyloxy)-2-methylbenzoic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: LHVJUPHCLWIPLZ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC(C(O)=O)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)