accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrBrN

CAS RN 1689-84-5

IUPAC Name: 3,5-dibromo-4-hydroxybenzonitrile
Synonyms: sabre caswell no. 119 brominal triple certrol B buctril industrial bromoxynil (substance) C7H3Br2NO brominal plus epa pesticide chemical code 035301 bromoxynil, sodium salt
Molecular Weight (g/mol): 276.92
Molecular Formula: C7H3Br2NO
InChi Key: UPMXNNIRAGDFEH-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(C=C1Br)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results