accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 16890-91-8

IUPAC Name: 1,4-dichlorocyclohexane
Synonyms: 1,4-dichlorocyclohexane
Molecular Weight (g/mol): 153.05
Molecular Formula: C6H10Cl2
InChi Key: WQTINZDWAXJLGH-UHFFFAOYSA-N
SMILES: ClC1CCC(Cl)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results