accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaFSiFFFFF

CAS RN 16893-85-9

IUPAC Name: disodium hexafluorosilanediuide
Synonyms: disodium hexafluorosilanediuide
Molecular Weight (g/mol): 188.06
Molecular Formula: F6Na2Si
InChi Key: TWGUZEUZLCYTCG-UHFFFAOYSA-N
SMILES: [Na+].[Na+].F[Si--](F)(F)(F)(F)F
Grade 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)