accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 168971-68-4

IUPAC Name: 1-bromo-2-fluoro-4-(trifluoromethoxy)benzene
Synonyms: 1-bromo-2-fluoro-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 259
Molecular Formula: C7H3BrF4O
InChi Key: KZMHSGBETSENAT-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results