CAS RN 1690-75-1
IUPAC Name:
methyl 1-methylpiperidine-4-carboxylate
Synonyms:
methyl 1-methylpiperidine-4-carboxylate
Molecular Weight (g/mol):
157.21
Molecular Formula:
C8H15NO2
InChi Key:
KYAOKPRJTMFBTQ-UHFFFAOYSA-N
SMILES:
COC(=O)C1CCN(C)CC1
1
–
1
of
1
results
1
Methyl 1-Methyl-4-piperidinecarboxylate 98.0+%, TCI America™
CAS: 1690-75-1 Molecular Formula: C8H15NO2 Molecular Weight (g/mol): 157.213 MDL Number: MFCD06658483 InChI Key: KYAOKPRJTMFBTQ-UHFFFAOYSA-N Synonym: 1-Methyl-4-piperidinecarboxylic Acid Methyl Ester PubChem CID: 3616844 IUPAC Name: methyl 1-methylpiperidine-4-carboxylate SMILES: CN1CCC(CC1)C(=O)OC