accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3OHO(E)

CAS RN 16909-11-8

IUPAC Name: (2E)-3-(3,5-dimethoxyphenyl)prop-2-enoic acid
Synonyms: (2E)-3-(3,5-dimethoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: VLSRUFWCGBMYDJ-ONEGZZNKSA-N
SMILES: COC1=CC(\C=C\C(O)=O)=CC(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results