accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4NH4PtClClClClClCl

CAS RN 16919-58-7

IUPAC Name: platinum(4+) diammonium hexachloride
Synonyms: platinum(4+) diammonium hexachloride
Molecular Weight (g/mol): 443.86
Molecular Formula: Cl6H8N2Pt
InChi Key: PCCGQTHFYHJATL-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)