CAS RN 16919-73-6
IUPAC Name:
dipotassium hexachloropalladiumdiuide
Synonyms:
dipotassium hexachloropalladate(2-)
Molecular Weight (g/mol):
403.36
Molecular Formula:
Cl6H6K2Pd
InChi Key:
XISJPQLCGXNMQD-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.Cl.Cl.Cl.[K+].[K+].[Pd--]
1
–
1
of
1
results
1
Sigma Aldrich Potassium hexachloropalladate(IV)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.