accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16919-74-7

IUPAC Name: dipotassium tetrabromopalladium dibromide
Synonyms: dipotassium tetrabromopalladium dibromide
Molecular Weight (g/mol): 664.04
Molecular Formula: Br6K2Pd
InChi Key: SKCNWISEAGYZFC-UHFFFAOYSA-H
SMILES: [K+].[K+].[Br-].[Br-].Br[Pd](Br)(Br)Br
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results