accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169205-78-1

IUPAC Name: N4-(3-bromophenyl)quinazoline-4,6-diamine
Synonyms: N4-(3-bromophenyl)quinazoline-4,6-diamine
Molecular Weight (g/mol): 315.17
Molecular Formula: C14H11BrN4
InChi Key: IZQHULBHKPGOAP-UHFFFAOYSA-N
SMILES: NC1=CC2=C(NC3=CC(Br)=CC=C3)N=CN=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results