accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16921-91-8

IUPAC Name: hexafluoro-λ⁵-phosphanuide; iminooxidanium
Synonyms: nitrosonium hexafluorophosphate
Molecular Weight (g/mol): 174.97
Molecular Formula: F6NOP
InChi Key: SAKPNYRUBHJGAR-UHFFFAOYSA-N
SMILES: N#[O+].F[P-](F)(F)(F)(F)F
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)